Assay Design



Gene Transcripts

Gene Symbol
FCRL4
Entrez Gene ID
102170192
Gene Name
Fc receptor like 4
Gene Aliases
-
Chromosome Location
Chr.3: 107264513 - 107288860 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013976239.2 XP_013831693.2 2 - 3 352 79
GenBank mRNA JT686153.1 - 3 - 4 335 79

View Full Product Details