Assay Design



Gene Transcripts

Gene Symbol
HSCB
Entrez Gene ID
102188337
Gene Name
HscB mitochondrial iron-sulfur cluster cochaperone
Gene Aliases
-
Chromosome Location
Chr.17: 3074643 - 3083642 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691708.3 XP_005691765.3 4 - 5 633 72
XM_018061049.1 XP_017916538.1 3 - 4 338 72
GenBank mRNA KA349629.1 - 4 - 5 629 72

View Full Product Details