Assay Design



Gene Transcripts

Gene Symbol
EMILIN3
Entrez Gene ID
102182704
Gene Name
elastin microfibril interfacer 3
Gene Aliases
-
Chromosome Location
Chr.13: 69553349 - 69559360 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057881.1 XP_017913370.1 2 - 3 293 96

View Full Product Details