Assay Design



Gene Transcripts

Gene Symbol
PGPEP1L
Entrez Gene ID
102182070
Gene Name
pyroglutamyl-peptidase I like
Gene Aliases
-
Chromosome Location
Chr.21: 7347027 - 7417292 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972902.2 XP_013828356.2 1 - 2 325 55

View Full Product Details