Assay Design



Gene Transcripts

Gene Symbol
MOAP1
Entrez Gene ID
102183731
Gene Name
modulator of apoptosis 1
Gene Aliases
-
Chromosome Location
Chr.21: 57276776 - 57278838 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065929.1 XP_017921418.1 2 - 3 834 81

View Full Product Details