Assay Design



Gene Transcripts

Gene Symbol
AZU1
Entrez Gene ID
102185651
Gene Name
azurocidin 1
Gene Aliases
-
Chromosome Location
Chr.7: 67088322 - 67091536 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682894.3 XP_005682951.1 1 - 2 345 64

View Full Product Details