Assay Design



Gene Transcripts

Gene Symbol
GINS2
Entrez Gene ID
102183625
Gene Name
GINS complex subunit 2
Gene Aliases
-
Chromosome Location
Chr.18: 13045844 - 13086703 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061811.1 XP_017917300.1 3 - 4 443 63
GenBank mRNA JO605686.2 - 3 - 4 384 63

View Full Product Details