Assay Design



Gene Transcripts

Gene Symbol
TAX1BP3
Entrez Gene ID
102187259
Gene Name
Tax1 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.19: 24166706 - 24172069 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693411.3 XP_005693468.1 3 - 4 430 81
GenBank mRNA KA345534.1 - 3 - 4 308 81

View Full Product Details