Assay Design



Gene Transcripts

Gene Symbol
SAMD10
Entrez Gene ID
102189598
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
-
Chromosome Location
Chr.13: 53193877 - 53198629 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057490.1 XP_017912979.1 2 - 3 572 53
XM_018057491.1 XP_017912980.1 2 - 3 572 53

View Full Product Details