Assay Design



Gene Transcripts

Gene Symbol
SMAGP
Entrez Gene ID
102188018
Gene Name
small cell adhesion glycoprotein
Gene Aliases
-
Chromosome Location
Chr.5: 28201040 - 28223308 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047755.1 XP_017903244.1 3 - 4 326 76
GenBank mRNA JO603280.2 - 1 - 2 149 76
KA318813.1 - 3 - 4 368 76

View Full Product Details