Assay Design



Gene Transcripts

Gene Symbol
CLEC12A
Entrez Gene ID
102187749
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
-
Chromosome Location
Chr.5: 98385927 - 98398661 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680858.1 XP_005680915.1 3 - 4 485 98
GenBank mRNA JO417360.2 - 3 - 4 339 98
JO612746.2 - 3 - 4 489 98

View Full Product Details