Assay Design



Gene Transcripts

Gene Symbol
UCMA
Entrez Gene ID
102173757
Gene Name
upper zone of growth plate and cartilage matrix associated
Gene Aliases
-
Chromosome Location
Chr.13: 27259552 - 27266600 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687951.3 XP_005688008.1 3 - 4 276 79
GenBank mRNA JO416756.2 - 3 - 4 240 79

View Full Product Details