Assay Design



Gene Transcripts

Gene Symbol
STMN2
Entrez Gene ID
102177815
Gene Name
stathmin 2
Gene Aliases
-
Chromosome Location
Chr.14: 39194911 - 39253320 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689118.2 XP_005689175.1 3 - 4 672 64
GenBank mRNA JO635168.2 - 2 - 3 1549 64
JT721282.1 - 4 - 5 547 64

View Full Product Details