Assay Design



Gene Transcripts

Gene Symbol
ST8SIA3
Entrez Gene ID
102169151
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.24: 56818810 - 56827518 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697280.3 XP_005697337.1 1 - 2 519 75
GenBank mRNA JO648240.2 - 1 - 2 385 75

View Full Product Details