Assay Design



Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
102175835
Gene Name
membrane spanning 4-domains A15
Gene Aliases
-
Chromosome Location
Chr.29: 36358472 - 36367446 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699651.2 XP_005699708.1 1 - 2 250 97
XM_005699653.2 XP_005699710.1 1 - 2 231 97

View Full Product Details