Assay Design



Gene Transcripts

Gene Symbol
SYNDIG1L
Entrez Gene ID
102189764
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
-
Chromosome Location
Chr.10: 17393009 - 17410762 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686075.3 XP_005686132.1 4 - 5 1106 75
GenBank mRNA JO644903.2 - 1 - 2 180 75

View Full Product Details