Assay Design
Gene Transcripts
- Gene Symbol
- TMEM91
- Entrez Gene ID
-
102191641
- Gene Name
- transmembrane protein 91
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 51451369 - 51456740 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO625529.2 |
- |
2 - 3 |
419 |
75 |
View Full Product Details