Assay Design



Gene Transcripts

Gene Symbol
LOC102188106
Entrez Gene ID
102188106
Gene Name
lipase member J
Gene Aliases
LIPJ
Chromosome Location
Chr.26: 41209421 - 41230805 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698181.2 XP_005698238.2 6 - 7 794 87

View Full Product Details