Assay Design



Gene Transcripts

Gene Symbol
LACTBL1
Entrez Gene ID
102187011
Gene Name
lactamase beta like 1
Gene Aliases
-
Chromosome Location
Chr.2: 6079779 - 6099173 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018067081.1 XP_017922570.1 1 - 2 145 93

View Full Product Details