Assay Design



Gene Transcripts

Gene Symbol
CDK3
Entrez Gene ID
102189157
Gene Name
cyclin dependent kinase 3
Gene Aliases
-
Chromosome Location
Chr.19: 54869266 - 54873100 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972144.2 XP_013827598.1 1 - 2 264 110
XM_018063773.1 XP_017919262.1 1 - 2 264 110

View Full Product Details