Assay Design



Gene Transcripts

Gene Symbol
C2H2orf80
Entrez Gene ID
102189721
Gene Name
chromosome 2 C2orf80 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 39463981 - 39489778 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676445.2 XP_005676502.2 6 - 7 528 60

View Full Product Details