Assay Design



Gene Transcripts

Gene Symbol
CA5A
Entrez Gene ID
102191071
Gene Name
carbonic anhydrase 5A
Gene Aliases
-
Chromosome Location
Chr.18: 14745214 - 14771478 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691856.3 XP_005691913.1 5 - 6 711 80
GenBank mRNA JO402918.2 - 3 - 4 409 80
JO402919.2 - 2 - 3 421 80

View Full Product Details