Assay Design



Gene Transcripts

Gene Symbol
HOXD9
Entrez Gene ID
102186531
Gene Name
homeobox D9
Gene Aliases
-
Chromosome Location
Chr.2: 115529360 - 115531989 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013968710.2 XP_013824164.2 1 - 2 1316 56
GenBank mRNA JO435547.2 - 1 - 2 448 56

View Full Product Details