Assay Design



Gene Transcripts

Gene Symbol
PRICKLE4
Entrez Gene ID
102174709
Gene Name
prickle planar cell polarity protein 4
Gene Aliases
-
Chromosome Location
Chr.23: 33206088 - 33212393 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038735.1 XP_017894224.1 1 - 2 643 70

View Full Product Details