Assay Design



Gene Transcripts

Gene Symbol
IP6K3
Entrez Gene ID
102185047
Gene Name
inositol hexakisphosphate kinase 3
Gene Aliases
-
Chromosome Location
Chr.23: 40811955 - 40836106 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039298.1 XP_017894787.1 5 - 6 1148 106
XM_018039299.1 XP_017894788.1 5 - 6 1148 106

View Full Product Details