Assay Design



Gene Transcripts

Gene Symbol
BCDIN3D
Entrez Gene ID
102172602
Gene Name
BCDIN3 domain containing RNA methyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 29691600 - 29698978 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679982.3 XP_005680039.2 1 - 2 649 94

View Full Product Details