Assay Design



Gene Transcripts

Gene Symbol
R3HDML
Entrez Gene ID
102169773
Gene Name
R3H domain containing like
Gene Aliases
-
Chromosome Location
Chr.13: 72303940 - 72313413 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057899.1 XP_017913388.1 3 - 4 512 62

View Full Product Details