Assay Design



Gene Transcripts

Gene Symbol
ODR4
Entrez Gene ID
102177359
Gene Name
odr-4 GPCR localization factor homolog
Gene Aliases
C16H1orf27
Chromosome Location
Chr.16: 66075166 - 66113691 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690983.3 XP_005691040.1 7 - 8 805 77
XM_005690986.2 XP_005691043.1 6 - 7 709 77
XM_005690987.2 XP_005691044.1 6 - 7 683 77
GenBank mRNA JT709495.1 - 7 - 8 977 77

View Full Product Details