Assay Design
Gene Transcripts
- Gene Symbol
- CES3
- Entrez Gene ID
-
102177362
- Gene Name
- carboxylesterase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 35924647 - 35937281 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO419999.2 |
- |
11 - 12 |
1553 |
60 |
JO431746.2 |
- |
11 - 12 |
1510 |
60 |
View Full Product Details