Assay Design



Gene Transcripts

Gene Symbol
CES3
Entrez Gene ID
102177362
Gene Name
carboxylesterase 3
Gene Aliases
-
Chromosome Location
Chr.18: 35924647 - 35937281 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692115.3 XP_005692172.2 7 - 8 970 70
GenBank mRNA JO419999.2 - 7 - 8 1034 70
JO431746.2 - 7 - 8 991 70

View Full Product Details