Assay Design



Gene Transcripts

Gene Symbol
PANK1
Entrez Gene ID
102170661
Gene Name
pantothenate kinase 1
Gene Aliases
-
Chromosome Location
Chr.26: 39998937 - 40110516 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698201.3 XP_005698258.1 3 - 4 806 78
XM_018041474.1 XP_017896963.1 3 - 4 1451 78
XM_018041475.1 XP_017896964.1 5 - 6 1212 78
XM_018041476.1 XP_017896965.1 5 - 6 1063 78
GenBank mRNA JO601071.2 - 5 - 6 1017 78

View Full Product Details