Assay Design



Gene Transcripts

Gene Symbol
KLF17
Entrez Gene ID
102180450
Gene Name
Kruppel like factor 17
Gene Aliases
-
Chromosome Location
Chr.3: 18818218 - 18836813 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013962854.2 XP_013818308.2 4 - 5 1350 58

View Full Product Details