Assay Design



Gene Transcripts

Gene Symbol
ANKRD60
Entrez Gene ID
102176804
Gene Name
ankyrin repeat domain 60
Gene Aliases
-
Chromosome Location
Chr.13: 57433909 - 57444225 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056864.1 XP_017912353.1 3 - 4 705 103

View Full Product Details