Assay Design



Gene Transcripts

Gene Symbol
NKAIN1
Entrez Gene ID
102173703
Gene Name
sodium/potassium transporting ATPase interacting 1
Gene Aliases
-
Chromosome Location
Chr.2: 13524052 - 13570057 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057341.1 XP_017912830.1 4 - 5 742 70
XM_018057348.1 XP_017912837.1 3 - 4 643 70
GenBank mRNA KA324102.1 - 3 - 4 359 70

View Full Product Details