Assay Design



Gene Transcripts

Gene Symbol
CACTIN
Entrez Gene ID
102177710
Gene Name
cactin, spliceosome C complex subunit
Gene Aliases
-
Chromosome Location
Chr.7: 90239896 - 90252115 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050832.1 XP_017906321.1 1 - 2 241 98

View Full Product Details