Assay Design



Gene Transcripts

Gene Symbol
MANBAL
Entrez Gene ID
102181877
Gene Name
mannosidase beta like
Gene Aliases
-
Chromosome Location
Chr.13: 65803118 - 65827733 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688534.3 XP_005688591.1 2 - 3 309 74
XM_005688535.3 XP_005688592.1 2 - 3 262 74
XM_005688536.3 XP_005688593.1 2 - 3 231 74
XM_005688537.3 XP_005688594.1 2 - 3 231 74

View Full Product Details