Assay Design



Gene Transcripts

Gene Symbol
CCDC97
Entrez Gene ID
102170452
Gene Name
coiled-coil domain containing 97
Gene Aliases
-
Chromosome Location
Chr.18: 51404120 - 51414646 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062458.1 XP_017917947.1 4 - 5 1072 89

View Full Product Details