Assay Design



Gene Transcripts

Gene Symbol
ZNF12
Entrez Gene ID
106503582
Gene Name
zinc finger protein 12
Gene Aliases
-
Chromosome Location
Chr.25: 39349714 - 39381632 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040408.1 XP_017895897.1 4 - 5 665 64

View Full Product Details