Assay Design



Gene Transcripts

Gene Symbol
SLC22A16
Entrez Gene ID
102177630
Gene Name
solute carrier family 22 member 16
Gene Aliases
-
Chromosome Location
Chr.9: 27064882 - 27098251 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013966433.2 XP_013821887.2 5 - 6 2178 56
XM_018053071.1 XP_017908560.1 6 - 7 1512 56

View Full Product Details