Assay Design



Gene Transcripts

Gene Symbol
ZSWIM7
Entrez Gene ID
102178855
Gene Name
zinc finger SWIM-type containing 7
Gene Aliases
-
Chromosome Location
Chr.19: 33153709 - 33163312 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064726.1 XP_017920215.1 2 - 3 151 65
GenBank mRNA JO447109.2 - 2 - 3 123 65

View Full Product Details