Assay Design



Gene Transcripts

Gene Symbol
ALG11
Entrez Gene ID
102190970
Gene Name
ALG11 alpha-1,2-mannosyltransferase
Gene Aliases
-
Chromosome Location
Chr.12: 65179502 - 65192217 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687472.3 XP_005687529.2 1 - 2 77 94
XM_018056701.1 XP_017912190.1 1 - 2 77 94
GenBank mRNA JO434055.2 - 1 - 2 102 94

View Full Product Details