Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005688773.3 | XP_005688830.3 | 4 - 5 | 805 | 87 |
XM_018058789.1 | XP_017914278.1 | 5 - 6 | 460 | 87 | |
XM_018058790.1 | XP_017914279.1 | 5 - 6 | 457 | 87 | |
XM_018058791.1 | XP_017914280.1 | 4 - 5 | 397 | 87 | |
XM_018058792.1 | XP_017914281.1 | 5 - 6 | 744 | 87 | |
XM_018058793.1 | XP_017914282.1 | 4 - 5 | 691 | 87 | |
XM_018058794.1 | XP_017914283.1 | 2 - 3 | 201 | 87 |