Assay Design



Gene Transcripts

Gene Symbol
IL24
Entrez Gene ID
102184925
Gene Name
interleukin 24
Gene Aliases
-
Chromosome Location
Chr.16: 4044993 - 4048949 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060104.1 XP_017915593.1 3 - 4 425 83

View Full Product Details