Assay Design



Gene Transcripts

Gene Symbol
NENF
Entrez Gene ID
102184646
Gene Name
neudesin neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.16: 69779746 - 69787077 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060749.1 XP_017916238.1 2 - 3 298 94
GenBank mRNA JO376089.2 - 2 - 3 174 94

View Full Product Details