Assay Design



Gene Transcripts

Gene Symbol
GLOD5
Entrez Gene ID
102183582
Gene Name
glyoxalase domain containing 5
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 21007079 - 21018170 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700810.1 XP_005700867.1 3 - 4 241 62

View Full Product Details