Assay Design



Gene Transcripts

Gene Symbol
C14H8orf33
Entrez Gene ID
102189327
Gene Name
chromosome 14 C8orf33 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 81632309 - 81634680 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013969044.2 XP_013824498.1 4 - 5 462 70
GenBank mRNA JO587245.2 - 3 - 4 274 70

View Full Product Details