Assay Design



Gene Transcripts

Gene Symbol
TM2D3
Entrez Gene ID
102184097
Gene Name
TM2 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.21: 28424911 - 28437514 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066496.1 XP_017921985.1 2 - 3 210 78
GenBank mRNA JO638732.2 - 2 - 3 235 78

View Full Product Details