Assay Design



Gene Transcripts

Gene Symbol
MED30
Entrez Gene ID
102183081
Gene Name
mediator complex subunit 30
Gene Aliases
-
Chromosome Location
Chr.14: 35275767 - 35299044 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689137.3 XP_005689194.2 3 - 4 603 71
XM_018058372.1 XP_017913861.1 2 - 3 443 71

View Full Product Details