Assay Design



Gene Transcripts

Gene Symbol
MPV17L
Entrez Gene ID
102173252
Gene Name
MPV17 mitochondrial inner membrane protein like
Gene Aliases
-
Chromosome Location
Chr.25: 13683852 - 13690407 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697521.3 XP_005697578.3 1 - 2 393 88

View Full Product Details