Assay Design



Gene Transcripts

Gene Symbol
CCDC189
Entrez Gene ID
102173147
Gene Name
coiled-coil domain containing 189
Gene Aliases
-
Chromosome Location
Chr.25: 26788904 - 26794876 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697710.3 XP_005697767.1 5 - 6 952 86

View Full Product Details